Learn More
protocadherin gamma subfamily B, 7, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PCDHGB7.
Brand: Abnova H00056099-A01.50uL
Description
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq
Sequence: TLDRETQSAHHLVLTALDGGDPPRSGTAQIRILVIDANDNPPVFSQDVYRVSLREDVPPGTSILRVKATDQDEGINSEITYSFFGVADKAQHVSpecifications
| protocadherin gamma subfamily B, 7 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PCDHGB7. | |
| PCDHGB7 | |
| ME6/PCDH-GAMMA-B7 | |
| Mouse | |
| 50 μL | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_018927 | |
| PCDHGB7 | |
| PCDHGB7 (NP_061750, 199 a.a. ∼ 291 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 56099 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.