Learn More
proprotein convertase subtilisin/kexin type 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human PCSK2 protein.
Brand: Abnova H00005126-D01P.100ug
Description
The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a proinsulin-processing enzyme that plays a key role in regulating insulin biosynthesis. It is also known to cleave proopiomelanocortin, proenkephalin, prodynorphin and proluteinizing-hormone-releasing hormone. The use of alternate polyadenylation sites has been found for this gene. [provided by RefSeq
Sequence: MKGGCVSQWKAAAGFLFCVMVFASAERPVFTNHFLVELHKGGEDKARQVAAEHGFGVRKLPFAEGLYHFYHNGLAKAKRRRSLHHKQQLERDPRVKMALQQEGFDRKKRGYRDINEIDINMNDPLFTKQWYLINTGQADGTPGLDLNVAEAWELGYTGKGVTIGIMDDGIDYLHPDLASNYNAEASYDFSSNDPYPYPRYTDDWFNSHGTRCAGEVSAAANNNICGVGVAYNSKVAGIRMLDQPFMTDIIEASSISHMPQLIDIYSASWGPTDNGKTVDGPRELTLQAMADGVNKGRGGKGSIYVWASGDGGSYDDCNCDGYASSMWTISINSAINDGRTALYDESCSSTLASTFSNGRKRNPEAGVATTDLYGNCTLRHSGTSAAAPEAAGVFALALEANLGLTWRDMQHLTVLTSKRNQLHDEVHQWRRNGVGLEFNHLFGYGVLDAGAMVKMAKDWKTVPERFHCVGGSVQDPEKIPSTGKLVLTLTTDACEGKENFVRYLEHVQAVITVNATRRGDLNINMTSPMGTKSILLSRRPRDDDSKVGFDKWPFMTTHTWGEDARGTWTLELGFVGSAPQKGVLKEWTLMLHGTQSAPYIDQVVRDYQSKLAMSKKEELEEELDEAVERSLKSILNKNSpecifications
| proprotein convertase subtilisin/kexin type 2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human PCSK2 protein. | |
| PCSK2 | |
| NEC2/PC2/SPC2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5126 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002594.2 | |
| PCSK2 | |
| PCSK2 (NP_002585.2, 1 a.a. ∼ 638 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.