Learn More
PCTAIRE protein kinase 1, Mouse, Clone: 2E5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PCTK1.
Brand: Abnova H00005127-M02.100ug
Description
The protein encoded by this gene belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. It may play a role in signal transduction cascades in terminally differentiated cells. This gene is thought to escape X inactivation. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq
Sequence: MDRMKKIKRQLSMTLRGGRGIDKTNGAPEQIGLDESGGGGGSDPGEAPTRAAPGELRSARGPLSSAPEIVHEDLKMGSDGSpecifications
| PCTAIRE protein kinase 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC001048 | |
| PCTK1 | |
| PCTK1 (AAH01048, 1 a.a. ∼ 80 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 2E5 | |
| Mouse monoclonal antibody raised against a partial recombinant PCTK1. | |
| PCTK1 | |
| FLJ16665/PCTAIRE/PCTAIRE1/PCTGAIRE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 5127 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.