Learn More
PDPK1 (pS241) Mouse, PE, Clone: J66-653.44.17, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560092
Description
The serine/threonine kinase 3- Phosphoinositide- Dependent Protein Kinase-1 (PDPK1, also known as PDK1) contributes to the activation of many important kinases in the insulin and IGF-1 signaling pathways. It acts downstream of phosphatidylinositol 3-kinase (PI3-kinase) to phosphorylate residues in the activation loops of many cellular kinases, including protein kinase B (PKB/Akt), PKC isoforms, p70 S6 kinase, and PDPK1 itself. The autophosphorylation of PDPK1 at serine 241 (S241) has recently been suggested to play a role in the regulation of PDPK1. It has been proposed that PDPK1 activity plays a key role in the regulation of various cellular events such as cell proliferation, differentiation, and apoptosis.
The J666-653.44.17 monoclonal antibody recognizes the phosphorylated S241 in the activation loop of human PDPK1. The orthologous phosphorylation site in mouse and rat PDPK1 is S244.
Specifications
| PDPK1 (pS241) | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Phosphorylated Human PDPK1 Peptide | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| J66-653.44.17 | |
| PDK1 (pS241), PKB Kinase (pS241) | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Mouse, Rat | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.