missing translation for 'onlineSavingsMsg'
Learn More

period homolog 2 (Drosophila), Mouse, Polyclonal Antibody, Abnova™

Product Code. 16189485
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16189485 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16189485 Supplier Abnova Supplier No. H00008864A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant PER2.

This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. Circadian expression in the suprachiasmatic nucleus continues in constant darkness, and a shift in the light/dark cycle evokes a proportional shift of gene expression in the suprachiasmatic nucleus. The specific function of this gene is not yet known. [provided by RefSeq

Sequence: MNGYAEFPPSPSNPTKEPVEPQPSQVPLQEDVDMSSGSSGHETNENCSTGRDSQGSDCDDSGKELGMLVEPPDARQSPDTFSLMMAKSEHNPSTSGCSSD

Specifications

Antigen period homolog 2 (Drosophila)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PER2.
Formulation 50% glycerol
Gene PER2
Gene Accession No. NM_022817
Gene Alias FASPS/KIAA0347
Gene Symbols PER2
Host Species Mouse
Immunogen PER2 (NP_073728, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8864
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.