missing translation for 'onlineSavingsMsg'
Learn More

PFKFB4 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA528648

Additional Details : Weight : 0.01000kg

Product Code. 13442767

  • € 472.00 / 100µL
Estimated Shipment: 18-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Recommended positive controls: Raji, mouse liver, rat liver. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (85%), Chicken (86%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PFKFB4 Polyclonal antibody specifically detects PFKFB4 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

PFKFB4
Polyclonal
Unconjugated
PFKFB4
6-biphosphatase-4; 6PF-2-K/Fru-2,6-P2ase 4; 6PF-2-K/Fru-2,6-P2ASE testis-type isozyme; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 4; bifunctional enzyme with kinase and biphosphatase activities; C230090D14; Fructose-2,6-bisphosphatase; PFK/FBPase 4; Pfkfb4
Rabbit
Antigen affinity chromatography
RUO
270198, 5210, 54283
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.73 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
P25114, Q16877, Q6DTY7
PFKFB4
Recombinant fragment corresponding to a region within amino acids 200 and 469 of Human PFKFB4
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers