missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ATRIP (Ser224) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539777
Additional Details : Weight : 0.00900kg
Description
ATRIP is an 86 kDa protein that is phosphorylated by ATR, regulates ATR expression, and is an essential component of the DNA damage checkpoint pathway. It localizes to the intranuclear foci after DNA damage or inhibition of replication.Specifications
Phospho-ATRIP (Ser224) | |
Polyclonal | |
Unconjugated | |
ATRIP | |
ATM and Rad3-related-interacting protein, ATR interacting protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84126 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8WXE1 | |
ATRIP | |
A synthetic phosphopeptide derived from human ATRIP around the phosphorylation site of Ser224 (H-V-SP-P-R) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |