Learn More
Phospho-PTEN (Ser370) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen 441060G
Additional Details : Weight : 0.50000kg
Product Code. 10424883
  • € 590.00 / Each
Estimated Shipment: 18-04-2024
to see stock.
Description
Description
Phospho-PTEN (Ser370) Polyclonal antibody specifically detects Phospho-PTEN (Ser370) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications
Specifications
Phospho-PTEN (Ser370)
Polyclonal
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
O08586, P60484
PTEN
The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human PTEN that contains serine 370. The sequence is conserved in mouse and rat.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PTEN
10q23del; 2310035O07Rik; A130070J02Rik; AI463227; B430203M17Rik; BZS; CWS1; DEC; GLM2; homolog of human mutated in multiple advanced cancers; MHAM; mitochondrial phosphatase and tensin protein alpha; mitochondrial PTENalpha; Mmac; MMAC1; MMAC1 phosphatase and tensin homolog deleted on chromosome 10; mutated in multiple advanced cancers 1; Phosphatase and tensin homolog; phosphatase and tensin homolog (mutated in multiple advanced cancers 1); phosphatase and tensin homolog deleted on chromosome ten; phosphatase and tensin-like protein; phosphatidylinositol 3; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; phosphatidylinositol-3; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; protein tyrosine phosphatase and tensin-like protein; PTEN; pten tumor suppressor; PTEN1; TEP1
Rabbit
Antigen affinity chromatography
RUO
19211, 50557, 5728
-20°C