missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT2 (Tyr689) Polyclonal Antibody, FITC, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA546888
Additional Details : Weight : 0.00010kg
Description
STAT2 is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. In response to interferon (IFN), this protein forms a complex with STAT1 and IFN regulatory factor family protein p48 (ISGF3G), in which this protein acts as a transactivator, but lacks the ability to bind DNA directly. Transcription adaptor P300/CBP (EP300/CREBBP) has been shown to interact specifically with this protein, which is thought to be involved in the process of blocking IFN-alpha response by adenovirus.Specifications
Phospho-STAT2 (Tyr689) | |
Polyclonal | |
PBS with 0.5% BSA and 0.09% sodium azide; pH 7.4 | |
P52630 | |
STAT2 | |
Phosphopeptide containing the human STAT2 Y689 site | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry | |
FITC | |
STAT2 | |
STAT2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6773 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |