missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Tyrosine Hydroxylase (Ser40) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 368600
Additional Details : Weight : 0.50000kg
Description
Phospho-Tyrosine Hydroxylase (Ser40) Polyclonal antibody specifically detects Phospho-Tyrosine Hydroxylase (Ser40) in Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
Phospho-Tyrosine Hydroxylase (Ser40) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P04177 | |
TH | |
Synthetic phospho-peptide derived from the rat tyrosine hydroxylase (TH) protein | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
TH | |
dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25085 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |