Learn More
anti-PHOX2A, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00000401-PW2.1Set
Product Code. 16181754
  • € 843.00 / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-PHOX2A (300μL), Antibody pair for WB: mouse purified polyclonal anti-PHOX2A (50μg)
Specifications
PHOX2A
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
ARIX/CFEOM2/FEOM2/MGC52227/NCAM2/PMX2A
Mouse, Rabbit
RUO
401
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
PHOX2A
PHOX2A
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-PHOX2A (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-PHOX2A (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.