Learn More
anti-PIAS2, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00009063-PW1.1Set
Product Code. 16182954
  • € 843.00 / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-PIAS2 (300μL), Antibody pair for WB: mouse purified polyclonal anti-PIAS2 (50μg)
Specifications
PIAS2
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
MGC102682/MIZ1/PIASX/PIASX-ALPHA/PIASX-BETA/SIZ2/ZMIZ4/miz
Mouse, Rabbit
RUO
9063
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
PIAS2
PIAS2
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-PIAS2 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-PIAS2 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.