Learn More
PKCα Mouse anti-Human, Alexa Fluor 488, Clone: 3/PKC, BD
Description
The Protein Kinase C (PKC) family of homologous serine/threonine protein kinases is involved in a number of processes, such as growth, differentiation, and cytokine secretion. At least eleven isozymes have been described. These proteins are products of multiple genes and alternative splicing. Conventional PKC (cPKC) subfamily members (α, β, and γ isoforms) consists of a single polypeptide chain containing four conserved regions (C) and five variable regions (V). The N-terminal half containing C1, C2, V1, and V2 constitutes the regulatory domain and interacts with the PKC activators Ca2+, phospholipid, diacylglycerol, or phorbol ester. However, the the C2-like domains of
novel PKC (nPKC) subfamily members (δ, ε, η, and θ isoforms) are Ca2+-independent. The atypical PKC (aPKC) subfamily members (ζ, ι,
and λ isoforms) lack the C2 domain and are unique in that their activity is independent of diacylglycerols and phorbol esters. They also lack one repeat of the cysteine-rich sequences that are conserved in cPKC and nPKC members. The C-terminal region of PKC contains the catalytic domain. The PKC pathway represents a major signal transduction system that is activated following ligand-stimulation of transmembrane receptors by hormones, neurotransmitters, and growth factors. PKCα regulates a wide variety of functions such as cellular growth, apoptosis, cardiomyocyte function, and brain cognitive functions. The 3/PKCα monoclonal antibody recognizes PKCα, regardless of phosphorylation status, and has been reported to crossreact with PKCβ.
Host Species: Mouse
Clone: 3
Isotype: IgG2b, κ
Species Reactivity: Human
Immunogen: Human PKCα aa. 270-427 Recombinant Protein
Intracellular Staining
Specifications
Specifications
| Antigen | PKCα |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 3/PKC |
| Conjugate | Alexa Fluor 488 |
| Description | Protein Kinase C α, PKC-α, PKC-A, PKCA, PRKCA |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human PKCα aa. 270-427 Recombinant Protein |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.