Learn More
PLK1 (pT210) Mouse, Unlabeled, Clone: K50-483, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558400
Description
Polo-like kinase (PLK1) is a serine/threonine kinase with structural similarities to Drosophila's Polo kinase and the Cdc5p of Saccharomyces cerevisiae. Like its invertebrate counterparts, PLK1 activity is required for DNA synthesis and is regulated throughout the cell cycle. Furthermore, PLK1 is highly expressed in primary tumors. It associates with the mitotic spindle during mitosis suggesting that, in addition to its role during S phase, PLK1 may play a role during chromosome segregation. This is consistent with its potential role in cancer development. Threonine 210 (T210) is one of the major phoshorylation sites in activated PLK1 obtained from human mitotic cells. The K50-483 monoclonal antibody recognizes the phosphorylated T210 of human PLK1.
Western Blotting
Specifications
| PLK1 (pT210) | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Phosphorylated Human PLK1 Peptide | |
| 0.1 mg | |
| Cell Biology | |
| Human, Mouse | |
| IgG1 κ |
| Western Blot | |
| K50-483 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.