Learn More
anti-POLR3K Matched Antibody Pair, Abnova
Brand: Abnova H00051728-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-POLR3K (100μg); Mouse polyclonal anti-POLR3K (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This gene encodes a small essential subunit of RNA polymerase III, the polymerase responsible for synthesizing transfer and small ribosomal RNAs in eukaryotes. The carboxy-terminal domain of this subunit shares a high degree of sequence similarity to the carboxy-terminal domain of an RNA polymerase II elongation factor. This similarity in sequence is supported by functional studies showing that this subunit is required for proper pausing and termination during transcription.
ELISA
Specifications
| POLR3K | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human POLR3K. | |
| C11/C11-RNP3/My010/RPC10/RPC11/hRPC11 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-POLR3K (100 μg) 2. Detection antibody: mouse polyclonal anti-POLR3K (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| POLR3K | |
| POLR3K | |
| Affinity Purified | |
| RUO | |
| 51728 | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.