Learn More
PPARGC1A, Mouse, Clone: 2G8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PPARGC1A.
Brand: Abnova H00010891-M17.100ug
Description
The protein encoded by this gene is a transcriptional coactivator that regulates the genes involved in energy metabolism. This protein interacts with PPARgamma, which permits the interaction of this protein with multiple transcription factors. This protein can interact with, and regulate the activities of, cAMP response element binding protein (CREB) and nuclear respiratory factors (NRFs). It provides a direct link between external physiological stimuli and the regulation of mitochondrial biogenesis, and is a major factor that regulates muscle fiber type determination. This protein may be also involved in controlling blood pressure, regulating cellular cholesterol homoeostasis, and the development of obesity. [provided by RefSeq
Sequence: TRTELRDRFEVFGEIEECTVNLRDDGDSYGFITYRYTCDAFAALENGYTLRRSNETDFELYFCGRKQFFKSNYADLDSNSDDFDPASTKSKYDSLDFDSLLKEAQRSLRRSpecifications
| PPARGC1A, | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| NM_013261 | |
| PPARGC1A | |
| PPARGC1A (NP_037393, 689 a.a. ∼ 798 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10891 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2G8 | |
| Mouse monoclonal antibody raised against a partial recombinant PPARGC1A. | |
| PPARGC1A | |
| LEM6/PGC-1(alpha)/PGC-1v/PGC1/PGC1A/PPARGC1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.