Learn More
protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PPFIA3.
Brand: Abnova H00008541-A01.50uL
Description
The protein encoded by this gene is a member of the LAR protein-tyrosine phosphatase-interacting protein (liprin) family. Liprins interact with members of LAR family of transmembrane protein tyrosine phosphatases, which are known to be important for axon guidance and mammary gland development. Liprin family protein has been shown to localize phosphatase LAR to cell focal adhesions and may be involved in the molecular organization of presynaptic active zones. [provided by RefSeq
Sequence: NERVMGWVSGLGLKEFATNLTESGVHGALLALDETFDYSDLALLLQIPTQNAQARQLLEKEFSNLISLGTDRRLDEDSAKSFSRSPSWRKMFREKDLSpecifications
| protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant PPFIA3. | |
| PPFIA3 | |
| KIAA0654/LPNA3/MGC126567/MGC126569 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 8541 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003660 | |
| PPFIA3 | |
| PPFIA3 (NP_003651, 1043 a.a. ∼ 1139 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.