Learn More
peroxiredoxin 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Description
This gene encodes a protein with antioxidant function and is localized in the mitochondrion. This gene shows significant nucleotide sequence similarity to the gene coding for the C22 subunit of Salmonella typhimurium alkylhydroperoxide reductase. Expression of this gene product in E. coli deficient in the C22-subunit gene rescued resistance of the bacteria to alkylhydroperoxide. The human and mouse genes are highly conserved, and they map to the regions syntenic between mouse and human chromosomes. Sequence comparisons with recently cloned mammalian homologues suggest that these genes consist of a family that is responsible for regulation of cellular proliferation, differentiation, and antioxidant functions. Two transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | peroxiredoxin 3 |
| Applications | Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against a full-length human PRDX3 protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PRDX3 |
| Gene Accession No. | NM_006793.2 |
| Gene Alias | AOP-1/AOP1/MER5/MGC104387/MGC24293/PRO1748/SP-22 |
| Gene Symbols | PRDX3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.