Learn More
prostaglandin D2 synthase 21kDa (brain), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PTGDS protein.
Brand: Abnova H00005730-B01.50uL
Description
The protein encoded by this gene is a glutathione-independent prostaglandin D synthase that catalyzes the conversion of prostaglandin H2 (PGH2) to postaglandin D2 (PGD2). PGD2 functions as a neuromodulator as well as a trophic factor in the central nervous system. PGD2 is also involved in smooth muscle contraction/relaxation and is a potent inhibitor of platelet aggregation. This gene is preferentially expressed in brain. Studies with transgenic mice overexpressing this gene suggest that this gene may be also involved in the regulation of non-rapid eye movement sleep. [provided by RefSeq
Sequence: MATHHTLWMGLALLGVLGDLQAAPEAQVSVQPNFQQDKFLGRWFSAGLASNSSWLREKKAALSMCKSVVAPATDGGLNLTSTFLRKNQCETRTMLLQPAGSLGSYSYRSPHWGSTYSVSVVETDYDQYALLYSQGSKGPGEDFRMATLYSRTQTPRAELKEKFTAFCKAQGFTEDTIVFLPQTDKCMTEQSpecifications
| prostaglandin D2 synthase 21kDa (brain) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human PTGDS protein. | |
| PTGDS | |
| LPGDS/PDS/PGD2/PGDS/PGDS2 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 5730 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_000954.5 | |
| PTGDS | |
| PTGDS (NP_000945.3, 1 a.a. ∼ 190 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.