Learn More
PTK7 protein tyrosine kinase 7, Mouse, Clone: 4F5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PTK7.
Brand: Abnova H00005754-M06.100ug
Description
Receptor protein tyrosine kinases transduce extracellular signals across the cell membrane. A subgroup of these kinases lack detectable catalytic tyrosine kinase activity but retain roles in signal transduction. The protein encoded by this gene is a member of this subgroup of tyrosine kinases and may function as a cell adhesion molecule. This gene is thought to be expressed in colon carcinomas but not in normal colon, and therefore may be a marker for or may be involved in tumor progression. Four transcript variants encoding four different isoforms have been found for this gene. [provided by RefSeq
Sequence: KQPSSQDALQGRRALLRCEVEAPGPVHVYWLLDGAPVQDTERRFAQGSSLSFAAVDRLQDSGTFQCVARDDVTGEEARSANASFNIKWIEAGPVVLKHPASEAEIQPQTQSpecifications
| PTK7 protein tyrosine kinase 7 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002821 | |
| PTK7 | |
| PTK7 (NP_002812, 36 a.a. ∼ 145 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 4F5 | |
| Mouse monoclonal antibody raised against a partial recombinant PTK7. | |
| PTK7 | |
| CCK4 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 5754 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.