missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB11B, member RAS oncogene family, Mouse, Clone: 2F1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RAB11B.
Brand: Abnova H00009230-M03A.200uL
This item is not returnable.
View return policy
Description
Sequence: MGTRDDEYDYLFKVVLIGDSGVGKSNLLSRFTRNEFNLESKSTIGVEFATRSIQVDGKTIKAQIWDTAGQERYRAITSAYYRGAVGALLVYDIAKHLTYENVERWLKELRSpecifications
| RAB11B, member RAS oncogene family | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_004218 | |
| RAB11B | |
| RAB11B (NP_004209, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Yes | |
| 9230 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA | |
| 2F1 | |
| Mouse monoclonal antibody raised against a partial recombinant RAB11B. | |
| RAB11B | |
| H-YPT3/MGC133246 | |
| Mouse | |
| 200 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction