missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB32, member RAS oncogene family (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RAB32.
Brand: Abnova H00010981-A01.50uL
This item is not returnable.
View return policy
Description
Small GTP-binding proteins of the RAB family, such as RAB32, play essential roles in vesicle and granule targeting (Bao et al., 2002 [PubMed 11784320]).[supplied by OMIM
Sequence: IPAVLLANKCDQNKDSSQSPSQVDQFCKEHGFAGWFETSAKDNINIEEAARFLVEKILVNHQSFPNEENDVDKIKLDQETLRAENKSQCCSpecifications
| RAB32, member RAS oncogene family | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RAB32. | |
| RAB32 | |
| RAB32 | |
| RAB32 (NP_006825, 136 a.a. ∼ 225 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 10981 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006834 | |
| Mouse | |
| 50 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction