missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB7B, member RAS oncogene family, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RAB7B protein.
Brand: Abnova H00338382-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MNPRKKVDLKLIIVGAIGVGKTSLLHQYVHKTFYEEYQTTLGASILSKIIILGDTTLKLQIWDTGGQERFRSMVSTFYKGSDGCILAFDVTDLESFEALDIWRGDVLAKIVPMEQSYPMVLLGNKIDLADRKVPQEVAQGWCREKDIPYFEVSAKNDINVVQAFEMLASRALSRYQSILENHLTESIKLSPDQSRSRCCSpecifications
| RAB7B, member RAS oncogene family | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human RAB7B protein. | |
| RAB7B | |
| MGC16212/MGC9726/RAB7 | |
| Mouse | |
| 50 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC017092 | |
| RAB7B | |
| RAB7B (AAH17092, 1 a.a. ∼ 199 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 338382 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction