Learn More
RANBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1080
Additional Details : Weight : 0.01000kg
Description
PA1-080 detects Ran binding protein 1 (RanBP1) from human and mouse cells. PA1-080 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽30 kDa protein representing RanBP1 from HeLa cell extract. The PA1-080 immunogen is a synthetic peptide corresponding to residues M(1) A A A K D T H E D H D T S T E N T(18) C of human RanBP1. The PA1-080 immunizing peptide (Cat. # PEP-128) is available for use in neutralization and control experiments.
RANBP1 Polyclonal antibody specifically detects RANBP1 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
RANBP1 | |
Polyclonal | |
Unconjugated | |
RANBP1 | |
HpaII tiny fragments locus 9A; hpaII tiny fragments locus 9a protein; Htf9a; Htf9-a; Htf9-a/RanBP1; RAN binding protein 1; ran-binding protein 1; RANBP1; RANG; Ran-specific GTPase-activating protein; testis secretory sperm-binding protein Li 221n | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19385, 5902 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P34022, P43487 | |
RANBP1 | |
Synthetic peptide corresponding to residues M(1) A A A K D T H E D H D T S T E N T(18) C of human RanBP1. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |