missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Rat CD43 (W3/13), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65237-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD43 antibody from Proteintech is a mouse monoclonal antibody generated with rat thymocyte membrane glycoproteins of rat CD43. This antibody recognizes rat antigen. The CD43 antibody has been validated for the following applications: FC analysis.Specifications
| CD43 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 24796 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| W3/13 | |
| Unconjugated | |
| Rat thymocyte membrane glycoproteins | |
| 500 μg | |
| Primary | |
| Rat | |
| Flow Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction