missing translation for 'onlineSavingsMsg'
Learn More

Anti-Rat CD45 (OX1), Proteintech

Mouse Monoclonal Antibody

€ 64.00 - € 198.00

Specifications

Antigen CD45
Clone OX1
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30233781
View Documents
Proteintech
65228-1-IG-500UG
500 μg
€ 198.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30236030
View Documents
Proteintech
65228-1-IG-100UG
100 μg
€ 64.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CD45 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of rat CD45. This antibody recognizes rat antigen. The CD45 antibody has been validated for the following applications: FC analysis.
Specifications

Specifications

CD45
0.5 mg/mL
Monoclonal
Liquid
RUO
24699
Primary
Store at 2-8°C. Stable for one year after shipment.
OX1
Flow Cytometry
Unconjugated
Mouse
Rat
IgG1 κ
Affinity purification
Flow Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.