missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Rat CD45RA (OX-33), Proteintech
Mouse Monoclonal Antibody
€ 64.42 - € 199.29
Specifications
| Antigen | CD45RA |
|---|---|
| Clone | OX-33 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234666
|
Proteintech
65234-1-IG-500UG |
500 μg |
€ 199.29
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235454
|
Proteintech
65234-1-IG-100UG |
100 μg |
€ 64.42
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD45RA antibody from Proteintech is a mouse monoclonal antibody generated with leukocyte common antigen purified from rat splenocytes of rat CD45RA. This antibody recognizes rat antigen. The CD45RA antibody has been validated for the following applications: FC analysis.Specifications
| CD45RA | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 24699 | |
| IgG1 κ | |
| Affinity purification | |
| Flow Antibody |
| OX-33 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Rat | |
| Leukocyte common antigen purified from rat splenocytes | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title