Learn More
regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RCBTB2.
Brand: Abnova H00001102-A01.50uL
Description
This gene encodes a member of the RCC1-related GEF family. The N-terminal half of the encoded amino acid sequence shows similarity to the regulator of chromosome condensation RCC1, which acts as a guanine nucleotide exchange factor (GEF) protein for the Ras-related GTPase Ran. [provided by RefSeq
Sequence: TIEPRRLDSLNGKKIACLSYGSGPHIVLATTEGEVFTWGHNAYSQLGNGTTNHGLVPCHISTNLSNKQVIEVACGSYHSLVLTSDGEVFAWGYNNSGQVGSGSTVNQPISpecifications
| regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RCBTB2. | |
| RCBTB2 | |
| CHC1L | |
| Mouse | |
| 50 μL | |
| Yes | |
| 1102 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001268 | |
| RCBTB2 | |
| RCBTB2 (NP_001259, 86 a.a. ∼ 194 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.