Learn More
RCVRN, Human, IP-WB Antibody Pair (PW5), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00005957-PW5.1Set
Product Code. 16162554
  • € 843.00 / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-RCVRN (300μL), Antibody pair for WB: mouse purified polyclonal anti-RCVRN (50μg)
Specifications
RCVRN
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
RCV1
Mouse, Rabbit
RUO
5957
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
RCVRN
RCVRN
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-RCVRN (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-RCVRN (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.