missing translation for 'onlineSavingsMsg'
Learn More

replication factor C (activator 1) 1, 145kDa, Mouse, Clone: 3F8, Abnova™

Mouse monoclonal antibody raised against a partial recombinant RFC1.

Brand:  Abnova H00005981-M01.100ug

Product Code. 16154275

  • € 349.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is the large subunit of replication factor C, which is a five subunit DNA polymerase accessory protein. Replication factor C is a DNA-dependent ATPase that is required for eukaryotic DNA replication and repair. The protein acts as an activator of DNA polymerases, binds to the 3' end of primers, and promotes coordinated synthesis of both strands. It also may have a role in telomere stability. [provided by RefSeq]

Sequence: MDIRKFFGVIPSGKKLVSETVKKNEKTKSDEETLKAKKGIKEIKVNSSRKEDDFKQKQPSKKKRIIYDSDSESEETLQVKNAKKPPEKLPVSSKPGKISRQDPVTYISET
Specifications

Specifications

replication factor C (activator 1) 1, 145kDa
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
BC051786
RFC1
RFC1 (AAH51786, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
ELISA, Western Blot
3F8
Mouse monoclonal antibody raised against a partial recombinant RFC1.
RFC1
A1/MGC51786/MHCBFB/PO-GA/RECC1/RFC/RFC140
Mouse
Affinity Purified
RUO
5981
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG3 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.