Learn More
regulator of G-protein signaling 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human RGS3 protein.
Brand: Abnova H00005998-D01P.100ug
Description
This gene encodes a member of the regulator of G-protein signaling (RGS) family. This protein is a GTP-ase activating protein which inhibits G-protein mediated signal transduction. The protein is largely cytosolic, but G-protein activation leads to translocation of this protein to the plasma membrane. A nuclear form of this protein has also been described, but its sequence has not been identified. Multiple alternatively spliced transcript variants have been described for this gene but the full-length nature of some transcripts is not yet known. [provided by RefSeq
Sequence: MKNKLGIFRRRNESPGAPPAGKADKMMKSFKPTSEEALKWGESLEKLLVHKYGLAVFQAFLRTEFSEENLEFWLACEDFKKVKSQSKMASKAKKIFAEYIAIQACKEVNLDSYTREHTKDNLQSVTRGCFDLAQKRIFGLMEKDSYPRFLRSDLYLDLINQKKMSPPLSpecifications
| regulator of G-protein signaling 3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human RGS3 protein. | |
| RGS3 | |
| C2PA/FLJ20370/FLJ31516/FLJ90496/PDZ-RGS3/RGP3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_134427 | |
| RGS3 | |
| RGS3 (NP_602299.1, 1 a.a. ∼ 168 a.a) full-length human protein. | |
| 100 μg | |
| Signal Transduction | |
| 5998 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.