Learn More
rhomboid, veinlet-like 2 (Drosophila), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RHBDL2 protein.
Brand: Abnova H00054933-B01P.50ug
Description
The protein encoded by this gene is a member of the rhomboid family of integral membrane proteins. This family contains proteins that are related to Drosophila rhomboid protein. Members of this family are found in both prokaryotes and eukaryotes and are thought to function as intramembrane serine proteases. The encoded protein is thought to release soluble growth factors by proteolytic cleavage of certain membrane-bound substrates, including ephrin B2 and ephrin B3. [provided by RefSeq
Sequence: MNLNMGREMKEELEEEEKMREDGGGKDRAKSKKVHRIVSKWMLPEKSRGTYLERANCFPPPVFIISISLAELAVFIYYAVWKPQKQWITLDTGILESPFIYSPEKREEAWRFISYMLVHAGSpecifications
| rhomboid, veinlet-like 2 (Drosophila) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human RHBDL2 protein. | |
| RHBDL2 | |
| MGC16997/RRP2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 54933 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC013103.1 | |
| RHBDL2 | |
| RHBDL2 (AAH13103.1, 1 a.a. ∼ 121 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.