missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOG, Mouse, Clone: 2E6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RHOG.
Brand: Abnova H00000391-M05.100ug
This item is not returnable.
View return policy
Description
ARHG is a member of the RAS superfamily of genes, which encode GTP-binding proteins that act in the pathway of signal transduction and play a key role in the regulation of cellular functions.[supplied by OMIM
Sequence: RHKWHPEVCHHCPDVPILLVGTKKDLRAQPDTLRRLKEQGQAPITPQQGQALAKQIHAVRYLECSALQQDGVKEVFAEAVRAVLNPTPIKRGRSCILLSpecifications
| RHOG | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001665 | |
| RHOG | |
| RHOG (NP_001656, 94 a.a. ∼ 191 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2E6 | |
| Mouse monoclonal antibody raised against a partial recombinant RHOG. | |
| RHOG | |
| ARHG/MGC125835/MGC125836 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 391 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction