missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ras-like without CAAX 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RIT1.
Brand: Abnova H00006016-A01.50uL
This item is not returnable.
View return policy
Description
RIT belongs to the RAS (HRAS; MIM 190020) subfamily of small GTPases (Hynds et al., 2003 [PubMed 12668729]).[supplied by OMIM
Sequence: RVRRTDDTPVVLVGNKSDLKQLRQVTKEEGLALAREFSCPFFETSAAYRYYIDDVFHALVREIRRKEKEAVLAMEKKSKPKNSVWKRLKSPFRKKKDSSpecifications
| Ras-like without CAAX 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RIT1. | |
| RIT1 | |
| MGC125864/MGC125865/RIBB/RIT/ROC1 | |
| Mouse | |
| 50 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006912 | |
| RIT1 | |
| RIT1 (NP_008843, 120 a.a. ∼ 217 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 6016 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction