Learn More
roundabout, axon guidance receptor, homolog 2 (Drosophila), Mouse, Clone: 4G5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ROBO2.
Brand: Abnova H00006092-M01.100ug
Description
This gene belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. The encoded protein is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq
Sequence: ATGDPLPVISWLKEGFTFPGRDPRATIQEQGTLQIKNLRISDTGTYTCVATSSSGETSWSAVLDVTESGATISKNYDLSDLPGPPSKPQVTDVTKNSVTLSpecifications
| roundabout, axon guidance receptor, homolog 2 (Drosophila) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002942 | |
| ROBO2 | |
| ROBO2 (NP_002933, 441 a.a. ∼ 540 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4G5 | |
| Mouse monoclonal antibody raised against a partial recombinant ROBO2. | |
| ROBO2 | |
| KIAA1568/SAX3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 6092 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.