Learn More
SDHB, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SDHB protein.
Brand: Abnova H00006390-D01P.100ug
Description
Complex II of the respiratory chain, which is specifically involved in the oxidation of succinate, carries electrons from FADH to CoQ. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. The iron-sulfur subunit is highly conserved and contains three cysteine-rich clusters which may comprise the iron-sulfur centers of the enzyme. Sporadic and familial mutations in this gene result in paragangliomas and pheochromocytoma, and support a link between mitochondrial dysfunction and tumorigenesis. [provided by RefSeq
Sequence: MAAVVALSLRRRLPATTLGGACLQASRGAQTAAATAPRIKKFAIYRWDPDKAGDKPHMQTYEVDLNKCGPMVLDALIKIKNEVDSTLTFRRSCREGICGSCAMNINGGNTLACTRRIDTNLNKVSKIYPLPHMYVIKDLVPDLSNFYAQYKSIEPYLKKKDESQEGKQQYLQSIEEREKLDGLYECILCACCSTSCPSYWWNGDKYLGPAVLMQAYRWMIDSRDDFTEERLAKLQDPFSLYRCHTIMNCTRTCPKGLNPGKAIAEIKKMMATYKEKKASVSpecifications
| SDHB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human SDHB protein. | |
| SDHB | |
| FLJ92337/IP/PGL4/SDH/SDH1/SDHIP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6390 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC007840.2 | |
| SDHB | |
| SDHB (NP_002991.2, 1 a.a. ∼ 280 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.