Learn More
SEMA4A, Human, IP-WB Antibody Pair (PW1), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00064218-PW1.1Set
Product Code. 16153624
  • € 843.00 / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-SEMA4A (300μL), Antibody pair for WB: mouse purified polyclonal anti-SEMA4A (50μg)
Specifications
SEMA4A
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
CORD10/FLJ12287/RP35/SEMAB/SEMB
Mouse, Rabbit
RUO
64218
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
SEMA4A
SEMA4A
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-SEMA4A (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-SEMA4A (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.