missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Albumin Chicken anti-Human, Biotin, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Brand: Invitrogen PA172057
This item is not returnable.
View return policy
Description
PA1-72057 detects human serum albumin in human samples. PA1-72057 has been successfully used in ELISA and western blot procedures. The PA1-72057 immunogen is from purified normal human serum albumin.
Albumin is a soluble, monomeric protein with a molecular weight of approximately 65-67 kDa that comprises about one-half of the blood serum protein. Human serum albumin (HAS) is the most abundant protein in human blood plasma, and has a serum half-life of approximately 20 days. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular, unglycosylated serum protein synthesized in the liver as pre-pro-albumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough `produce the secreted albumin. Serum albumin has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. The main function of albumin is the regulation of the colloidal osmotic pressure of blood.
Specifications
| Human Serum Albumin | |
| Polyclonal | |
| Biotin | |
| P02768 | |
| ALB | |
| Purified normal human serum albumin. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgY |
| ELISA, Western Blot | |
| 1 mg/mL | |
| ALB | |
| Albumin, HSA, Serum albumin precursor, ALB | |
| Chicken | |
| Purified | |
| RUO | |
| 213 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction