missing translation for 'onlineSavingsMsg'
Learn More
Learn More
solute carrier family 22 (organic cation transporter), member 18 antisense, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLC22A18AS.
Brand: Abnova H00005003-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: EVVYSVPDNVPGQNGSRRPLVCKITGKCLSVCSEENAKAGGCSAFPLLLSQLGARMTGREHAHKGPELTTPDSGLPRPPNPALAGFRALAQHSPPLGTSTPSAVLLSAATSpecifications
| solute carrier family 22 (organic cation transporter), member 18 antisense | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SLC22A18AS. | |
| SLC22A18AS | |
| BWR1B/BWSCR1B/ORCTL2S/SLC22A1LS/p27-BWR1B | |
| Mouse | |
| 50 μL | |
| Yes | |
| 5003 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_007105 | |
| SLC22A18AS | |
| SLC22A18AS (NP_009036, 144 a.a. ∼ 253 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction