Learn More
SLC26A2, Mouse, Clone: 3F6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SLC26A2.
Brand: Abnova H00001836-M04.100ug
Description
The diastrophic dysplasia sulfate transporter is a transmembrane glycoprotein implicated in the pathogenesis of several human chondrodysplasias. It apparently is critical in cartilage for sulfation of proteoglycans and matrix organization. [provided by RefSeq
Sequence: MSSESKEQHNVSPRDSAEGNDSYPSGIHLELQRESSTDFKQFETNDQCRPYHRILIERQEKSDTNFKEFVIKKLQKNCQCSPAKAKNMILGFLPVLQWLPKYDLKKNISpecifications
| SLC26A2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_000112 | |
| SLC26A2 | |
| SLC26A2 (NP_000103.1, 1 a.a. ∼ 108 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| 3F6 | |
| Mouse monoclonal antibody raised against a partial recombinant SLC26A2. | |
| SLC26A2 | |
| D5S1708/DTD/DTDST/EDM4/MST153/MSTP157 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1836 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.