missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD family member 4, Mouse, Clone: 3E2, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SMAD4.
Brand: Abnova H00004089-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: MDNMSITNTPTSNDACLSIVHSLMCHRQGGESETFAKRAIESLVKKLKEKKDELDSLITAITTSGAHPSKCVTIQRTLDGRLQVAGRKGFPHVIYARLWRWPDLHKNELKHVKYCQYAFDLKCDSVCVNPYHYERVVSPGIDLSGLTLQSNAPSSMMVKDEYVHDFEGQPSLSTEGHSIQTIQHPPSNRASTETYSTPALLAPSESNATSTANFPNIPVASTSQPASILGGSHSEGLLQIASGPQPGQQQNGFTGQPATYHHNSTTTWTGSRTAPYTPNLPHHQNGHLQHHPPMPPHPGHYWPVHNELAFQPPISNHPAPEYWCSIAYFEMDVQVGETFKVPSSCPIVTVDGYVDPSGGDRFCLGQLSNVHRTEAIERARLHIGKGVQLECKGEGDVWVRCLSDHAVFVQSYYLDREAGRAPGDAVHKIYPSAYIKVFDLRQCHRQMQQQAATAQAAAAAQAAAVAGNIPGPGSVGGIAPAISLSAAAGIGVDDLRRLCILRMSFVKGWGPDYPRQSIKETPCWIEIHLHRALQLLDEVLHTMPIADPQPLDSpecifications
| SMAD family member 4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC002379 | |
| SMAD4 | |
| SMAD4 (AAH02379, 1 a.a. ∼ 552 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Signal Transduction | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, In situ PLA, Western Blot | |
| 3E2 | |
| Mouse monoclonal antibody raised against a full length recombinant SMAD4. | |
| SMAD4 | |
| DPC4/JIP/MADH4 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 4089 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction