Learn More
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1, Mouse, Clone: 3E3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SMARCB1.
Brand: Abnova H00006598-M02.100ug
Description
The protein encoded by this gene is part of a complex that relieves repressive chromatin structures, allowing the transcriptional machinery to access its targets more effectively. The encoded nuclear protein may also bind to and enhance the DNA joining activity of HIV-1 integrase. This gene has been found to be a tumor suppressor, and mutations in it have been associated with malignant rhabdoid tumors. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: YTTLATSVTLLKASEVEEILDGNDEKYKAVSISTEPPTYLREQKAKRNSQWVPTLPNSSHHLDAVPCSTTINRNRMGRDKKRTFPLCFDDHDPAVIHENASpecifications
| SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_003073 | |
| SMARCB1 | |
| SMARCB1 (NP_003064, 81 a.a. ∼ 180 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 3E3 | |
| Mouse monoclonal antibody raised against a partial recombinant SMARCB1. | |
| SMARCB1 | |
| BAF47/INI1/RDT/SNF5/SNF5L1/Sfh1p/Snr1/hSNFS | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 6598 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.