Learn More
sorting nexin 14, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SNX14.
Brand: Abnova H00057231-A01.50uL
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. The encoded protein also contains a regulator of G protein signaling (RGS) domain. Regulator of G protein signaling family members are regulatory molecules that act as GTPase activating proteins for G alpha subunits of heterotrimeric G proteins. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: QLFQEHRLVSLITLLRDAIFCENTEPRSLQDKQKGAKQTFEEMMNYIPDLLVKCIGEETKYESIRLLFDGLQQPVLNKQLTYVLLDIVIQELFPELNKVQKEVTSVTSWMSpecifications
| sorting nexin 14 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SNX14. | |
| SNX14 | |
| MGC13217/RGS-PX2 | |
| Mouse | |
| 50 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC005110 | |
| SNX14 | |
| SNX14 (AAH05110, 784 a.a. ∼ 893 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 57231 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.