Learn More
SRY (sex determining region Y)-box 12, Mouse, Clone: 2A6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SOX12.
Brand: Abnova H00006666-M06.100ug
Description
Members of the SOX family of transcription factors are characterized by the presence of a DNA-binding high mobility group (HMG) domain, homologous to the HMG box of sex-determining region Y (SRY). Forming a subgroup of the HMG domain superfamily, SOX proteins have been implicated in cell fate decisions in a diverse range of developmental processes. SOX transcription factors have diverse tissue-specific expression patterns during early development and have been proposed to act as target-specific transcription factors and/or as chromatin structure regulatory elements. The protein encoded by this gene was identified as a SOX family member based on conserved domains and its expression in various tissues suggests a role in both differentiation and maintenance of several cell types. [provided by RefSeq
Sequence: LGFLSRLPPGPAGLDCSALDRDPDLQPPSGTSHFEFPDYCTPEVTEMIAGDWRPSSIADLVFSpecifications
| SRY (sex determining region Y)-box 12 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006943 | |
| SOX12 | |
| SOX12 (NP_008874, 252 a.a. ∼ 313 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2A6 | |
| Mouse monoclonal antibody raised against a partial recombinant SOX12. | |
| SOX12 | |
| SOX22 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 6666 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.