missing translation for 'onlineSavingsMsg'
Learn More

Sp3 transcription factor, Mouse, Clone: 2F6, Abnova™

Mouse monoclonal antibody raised against a full-length recombinant SP3.

Brand:  Abnova H00006670-M08.100ug

Product Code. 16125735

  • € 349.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene belongs to a family of Sp1 related genes that encode transcription factors that regulate transcription by binding to consensus GC- and GT-box regulatory elements in target genes. This protein contains a zinc finger DNA-binding domain and several transactivation domains, and has been reported to function as a bifunctional transcription factor that either stimulates or represses the transcription of numerous genes. Transcript variants encoding different isoforms have been described for this gene, and one has been reported to initiate translation from a non-AUG (AUA) start codon. Additional isoforms, resulting from the use of alternate downstream translation initiation sites, have also been noted. [provided by RefSeq

Sequence: TAGINADGHLINTGQAMDSSDNSERTGERVSPDINETNTDTDLFVPTSSSSQLPVTIDSTGILQQNTNSLTTSSGQVHSSDLQGNYIQSPVSEETQAQNIQVSTAQPVVQHLQLQESQQPTSQAQIVQGITPQTIHGVQASGQN*
Specifications

Specifications

Sp3 transcription factor
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_003111
SP3
SP3 (NP_003102, 287 a.a. ∼ 430 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Transcription Regulation
Primary
Human
Antibody
ELISA, Western Blot
2F6
Mouse monoclonal antibody raised against a full length recombinant SP3.
SP3
DKFZp686O1631/SPR-2
Mouse
Affinity Purified
RUO
Yes
6670
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.