missing translation for 'onlineSavingsMsg'
Learn More

anti-Streptavidin, Brilliant Violet 786, Clone: Streptavidin, BD

Brand:  BD Biosciences 563858

Product Code. 15853159

  • ONLINE EXCLUSIVE PRICE
    € 720.00
    € 699.08 / Each
    Save € 20.92
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Host Species:
Clone: Streptavidin
Species Reactivity: Ms
Immunogen: Streptavidin from Streptomyces avidinii

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Streptavidin
Streptavidin
Brilliant Violet 786
Affinity Purified
RUO
Mouse
Monoclonal
0.1mg/mL
Streptavidin from Streptomyces avidinii
100 μg
Primary
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.