Learn More
synapsin I, Mouse, Clone: 4H1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SYN1.
Brand: Abnova H00006853-M07.100ug
Description
This gene is a member of the synapsin gene family. Synapsins encode neuronal phosphoproteins which associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains, and they are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. This member of the synapsin family plays a role in regulation of axonogenesis and synaptogenesis. The protein encoded serves as a substrate for several different protein kinases and phosphorylation may function in the regulation of this protein in the nerve terminal. Mutations in this gene may be associated with X-linked disorders with primary neuronal degeneration such as Rett syndrome. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: EIFGGLDICAVEALHGKDGRDHIIEVVGSSMPLIGDHQDEDKQLIVELVVNKMAQALPRQRQRDASPGRGSHGQTPSPGALPLGRQTSQSpecifications
| synapsin I | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006950 | |
| SYN1 | |
| SYN1 (NP_008881, 362 a.a. ∼ 450 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 6853 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4H1 | |
| Mouse monoclonal antibody raised against a partial recombinant SYN1. | |
| SYN1 | |
| SYN1a/SYN1b/SYNI | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.