Learn More
Synphilin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA118270
Additional Details : Weight : 0.01000kg
Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
Synphilin 1 Polyclonal antibody specifically detects Synphilin 1 in Human, Rat samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
Synphilin 1 | |
Polyclonal | |
Unconjugated | |
SNCAIP | |
2810407O15Rik; 4933427B05Rik; Alpha-synuclein-interacting protein; BB104594; MGC39814; SNCAIP; Sph1; Synphilin 1; synphilin-1; synuclein alpha interacting protein; synuclein, alpha interacting protein; synuclein, alpha interacting protein (synphilin); SYPH1 | |
Rabbit | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Frozen), Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
Q9Y6H5 | |
SNCAIP | |
Synthetic peptide corresponding to residues C A S K G K N K A A of human synphilin-1a conjugated to KLH. | |
RUO | |
307309, 9627 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |