Learn More
TACR1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant TACR1.
Brand: Abnova H00006869-A01.50uL
Description
This gene belongs to a gene family of tachykinin receptors. These tachykinin receptors are characterized by interactions with G proteins and contain seven hydrophobic transmembrane regions. This gene encodes the receptor for the tachykinin substance P, also referred to as neurokinin 1. The encoded protein is also involved in the mediation of phosphatidylinositol metabolism of substance P. [provided by RefSeq
Sequence: PRLSATATKVVICVIWVLALLLAFPQGYYSTTETMPSRVVCMIEWPEHPNKIYEKVYHICVTVLIYFLPLLVIGYAYTVVGITLWASEIPGDSSDRYHEQVSAKSpecifications
| TACR1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant TACR1. | |
| TACR1 | |
| NK1R/NKIR/SPR/TAC1R | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001058 | |
| TACR1 | |
| TACR1 (NP_001049, 140 a.a. ∼ 243 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 6869 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.