Learn More
anti-TCR PAN gamma/delta , Clone: IMMU510, Beckman Coulter™
Description
The IMMU510 monoclonal antibody recognizes all gamma/delta T cells, regardless of their variable chains. It stains 1.6% to 8.9% of peripheral CD3+ cells from healthy donors. In Western blot, it recognizes the delta chain in reducing conditions. It immunoprecipitates the three isoforms of the gamma/delta T cell receptor. This antibody is suitable for immunohistochemistry on frozen sections. It has been used to study the distribution of gamma/delta T cells and their functions.
bold (begin)Host Species:bold (end) Mouse
bold (begin)Clone:bold (end) IMMU510
bold (begin)Isotype:bold (end) IgG1
bold (begin)Cross Reactivity:bold (end) Human
Enzyme Immunoassay, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | TCR Pan γ/δ |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 0.1 mg |
| Primary or Secondary | Primary |
| Target Species | Human |
| Form | Purified |
| Isotype | IgG1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.